Learn More
Grainger Roller Guide Bottom 3-way 17 5/8 in
Catalog No. 502767068
Encompass_Preferred
The Coxreels 1125 Series Roller Guide is a replacement part for 1125 Series hose reels. It is made of durable molded plastic with stainless steel axle pins and measures approximately 3 inches wide. The roller guide allows the hose to wind smoothly onto the reel drum without binding or kinking. This USA-made replacement part is easy to install by snapping into place. It is recommended for industrial, commercial, and residential applications using 1125 Series hose reels from Coxreels.
Catalog No. 50-276-7068 Supplier Grainger Supplier No. 16X578
The Coxreels 1125 Series Roller Guide is a replacement part for 1125 Series hose reels. It is made of durable molded plastic with stainless steel axle pins and measures approximately 3 inches wide. The roller guide allows the hose to wind smoothly onto the reel drum without binding or kinking. This USA-made replacement part is easy to install by snapping into place. It is recommended for industrial, commercial, and residential applications using 1125 Series hose reels from Coxreels.