Learn More
Grainger Roller Guide Bottom 3-way 17 5/8 in
Catalog No. 502765636
Encompass_Preferred
The Grainger 3-Way Roller Guide Bottom is a heavy-duty roller guide made of steel with a durable powder coat finish. It has 3 sealed precision ball bearing rollers and measures 17 5/8" long x 2 3/4" wide x 2 1/4" high. The rollers have a load capacity of 250 lbs each. This guide can be mounted to the bottom of a sliding door or drawer to provide smooth rolling action. It features adjustable mounting brackets and holes for easy installation. The 3-way design allows the door or drawer to move side to side as well as in and out.
Catalog No. 50-276-5636 Supplier Grainger Supplier No. 16X579
The Grainger 3-Way Roller Guide Bottom is a heavy-duty roller guide made of steel with a durable powder coat finish. It has 3 sealed precision ball bearing rollers and measures 17 5/8" long x 2 3/4" wide x 2 1/4" high. The rollers have a load capacity of 250 lbs each. This guide can be mounted to the bottom of a sliding door or drawer to provide smooth rolling action. It features adjustable mounting brackets and holes for easy installation. The 3-way design allows the door or drawer to move side to side as well as in and out.