Learn More
Grainger Roller Guide Top 3-way 17 5/8 in
Catalog No. 502765635
Encompass_Preferred
The Coxreels 1125 Series Roller Guide is a replacement part for 1125 Series hose reels. It is constructed of durable molded polymer with stainless steel axle pins and measures approximately 3 inches wide. The roller guide allows the hose to smoothly pay in and out while preventing hose abrasion. It is recommended for use with 1/2", 5/8", and 3/4" ID hose. This USA-made replacement part is easy to install and maintains performance for the life of the hose reel.
Catalog No. 50-276-5635 Supplier Grainger Supplier No. 16X576
The Coxreels 1125 Series Roller Guide is a replacement part for 1125 Series hose reels. It is constructed of durable molded polymer with stainless steel axle pins and measures approximately 3 inches wide. The roller guide allows the hose to smoothly pay in and out while preventing hose abrasion. It is recommended for use with 1/2", 5/8", and 3/4" ID hose. This USA-made replacement part is easy to install and maintains performance for the life of the hose reel.