Learn More
Grainger Roller Guide Top 3-way 24 in
Catalog No. 502768191
Encompass_Preferred
The Grainger 16X581 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-scratching fibers for cleaning and scouring. It has a 6 inch by 9 inch size and comes in a pack of 10 pads. The pads have level 1 cleaning power that can tackle light duty scrubbing jobs. They are disposable and ideal for cleaning pots, pans, dishes, counters, appliances, tools, equipment and more. The open web design allows debris to move through the pad during scrubbing.
Catalog No. 50-276-8191 Supplier Grainger Supplier No. 16X581
The Grainger 16X581 is a 3M Scotch-Brite General Purpose Hand Pad that features an open web construction with non-scratching fibers for cleaning and scouring. It has a 6 inch by 9 inch size and comes in a pack of 10 pads. The pads have level 1 cleaning power that can tackle light duty scrubbing jobs. They are disposable and ideal for cleaning pots, pans, dishes, counters, appliances, tools, equipment and more. The open web design allows debris to move through the pad during scrubbing.