Learn More
Grainger RIDGID Roller Pins, For Pipe Threading Machines, 2/Pk.
Catalog No. 502794750
Encompass_Preferred
These roller pins are essential replacement components designed for maintenance and repair of select threading and cutting equipment. They provide a precise fit to ensure optimal performance and tool longevity during heavy-duty pipe threading and cutting operations.
  • Package of 2 roller pins
  • Compatible with model 354/732 cutters
  • Suitable for use with 1210, 1215, 1233, and 1822-I pipe and bolt threading machines
  • Constructed for durable, reliable operation
Catalog No. 50-279-4750 Supplier Grainger Supplier No. 19V327
These roller pins are essential replacement components designed for maintenance and repair of select threading and cutting equipment. They provide a precise fit to ensure optimal performance and tool longevity during heavy-duty pipe threading and cutting operations.
  • Package of 2 roller pins
  • Compatible with model 354/732 cutters
  • Suitable for use with 1210, 1215, 1233, and 1822-I pipe and bolt threading machines
  • Constructed for durable, reliable operation
Product Title
Select an issue
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.