Learn More
Cole-Parmer Argos Technologies CELLroll CELLroll System, Culture Bottle/Tube Roller, 0.1 to 2.0 rpm; 90-260 VAC
Catalog No. 501960338 Shop All Cole-Parmer Products
Encompass_Preferred
CELLroll systems are programmable, modular roller systems that accommodate roller bottles with culture volumes from 300 to 400 mL. Systems operate by remote control and feature two ranges of rotating speeds. The control panel features a digital display, timer, and alarm; program for direction of rotation, bottle angle, velocity, and interval cycles. Systems are CO2 and moisture resistant. Systems include two decks that hold two roller bottles and can be expanded to accommodate up to 16 bottles (total of eight decks). Fits vertically or horizontally inside a CO2 incubator. Control unit can be used outside the incubator for simple and flexible programming. One deck measures 11-5/8"L x14-3/16"W x 9-¼"H.
Catalog No. 50-196-0338 Supplier Cole-Parmer Supplier No. 1305000
Add to Cart
Add to Cart
Request a Quote
CELLroll systems are programmable, modular roller systems that accommodate roller bottles with culture volumes from 300 to 400 mL. Systems operate by remote control and feature two ranges of rotating speeds. The control panel features a digital display, timer, and alarm; program for direction of rotation, bottle angle, velocity, and interval cycles. Systems are CO2 and moisture resistant. Systems include two decks that hold two roller bottles and can be expanded to accommodate up to 16 bottles (total of eight decks). Fits vertically or horizontally inside a CO2 incubator. Control unit can be used outside the incubator for simple and flexible programming. One deck measures 11-5/8"L x14-3/16"W x 9-¼"H.