Learn More
MSC DBI-SALA Rollgliss R550 Rescue and Descent Device, 500 ft., Yellow, Aluminum, 1 ea.
Catalog No. 5000688892
Encompass
The Rollgliss™ R550 is a versatile rescue and descent device designed for vertical evacuation and rescue operations. Supporting up to two users, this system provides automatic controlled descent with the flexibility of coworker-assisted or user-controlled descent capabilities.
  • Aluminium and stainless steel construction provides corrosion resistance for extended service life
  • Integrated rope ears enable greater descent speed control during assisted rescues
  • Dual brake design offers automatic descent with enhanced security
  • Integrated RFID tag for easy equipment inspection and tracking
  • Compatible with 9.5 mm (3/8 in.) kernmantle rope
  • Certified to OSHA, EN341, EN1496, and ANSI standards
Catalog No. 50-006-88892 Supplier MSC Supplier No. 20759643
The Rollgliss™ R550 is a versatile rescue and descent device designed for vertical evacuation and rescue operations. Supporting up to two users, this system provides automatic controlled descent with the flexibility of coworker-assisted or user-controlled descent capabilities.
  • Aluminium and stainless steel construction provides corrosion resistance for extended service life
  • Integrated rope ears enable greater descent speed control during assisted rescues
  • Dual brake design offers automatic descent with enhanced security
  • Integrated RFID tag for easy equipment inspection and tracking
  • Compatible with 9.5 mm (3/8 in.) kernmantle rope
  • Certified to OSHA, EN341, EN1496, and ANSI standards