Learn More
Ethos Biosciences Classic Wright-Giemsa Stain, 1gal, Ethos
Catalog No. 5000574190
Encompass_Preferred
Classical Wright–Giemsa Stain is a Romanowsky-type stain formulated for differential staining of peripheral blood smears, bone marrow specimens, and other cytologic preparations. The combined Wright and Giemsa dyes provide enhanced nuclear detail, cytoplasmic contrast, and reliable differentiation of leukocyte populations, erythrocytes, platelets, and parasites. Suitable for hematology, cytology, microbiology, and research laboratories, this stain supports manual differentials, morphology studies, and teaching applications. Compatible with standard Wright–Giemsa staining protocols and automated or manual systems, it delivers consistent, reproducible color results for routine and specialized laboratory use.
Catalog No. 50-005-74190 Supplier Ethos Biosciences Supplier No. 1742G
Classical Wright–Giemsa Stain is a Romanowsky-type stain formulated for differential staining of peripheral blood smears, bone marrow specimens, and other cytologic preparations. The combined Wright and Giemsa dyes provide enhanced nuclear detail, cytoplasmic contrast, and reliable differentiation of leukocyte populations, erythrocytes, platelets, and parasites. Suitable for hematology, cytology, microbiology, and research laboratories, this stain supports manual differentials, morphology studies, and teaching applications. Compatible with standard Wright–Giemsa staining protocols and automated or manual systems, it delivers consistent, reproducible color results for routine and specialized laboratory use.