Learn More
Selleck Chemical LLC Romidepsin (FK228, Depsipeptide) 5mg 128517-07-7 FR 901228, NSC 630176
SDP
Supplier:  Selleck Chemical LLC S30205MG
Encompass_Preferred
Romidepsin (FK228, Depsipeptide, FR 901228, NSC 630176) is a potent HDAC1 and HDAC2 inhibitor with IC50 of 36 nM and 47 nM in cell-free assays, respectively. Romidepsin (FK228/depsipeptide) controls growth and induces apoptosis in neuroblastoma tumor cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6238