Learn More
Apexbio Technology LLC Romidepsin (FK228, Depsipeptide), 128517-07-7, MFCD18433404, 1 mg
SDP
Supplier:  Apexbio Technology LLC A81731
Encompass_Preferred
MF: C24H36N4O6S2, MW: 540.7, Synonyms: Istodax, Antibiotic FR 901228, FK228, FR 901228, FK-228, Romidepsin. Solubility: 27.035mg/mL in DMSO, 35.27 mg/mL in EtOH with ultrasonic,insoluble in H2O. Romidepsin, also known as FK228 or depsipeptide, is potent and selective inhibitor of histone deacetylases (HDACs) which are associated with the regulation of re-expression of silenced tumor suppressor genes.
Catalog No. 50-101-4336