Learn More
Cayman Chemical RomIfIdInhydrochlorIde 50mg
SDP
Catalog No. 502910072
Encompass_Preferred
An α2-AR agonist; epidural administration induces antinociception and sedation in donkeys at 50 UG/kg; induces bradycardia in horses from 0.04-0.12 mg/kg
Catalog No. 50-291-0072 Supplier Cayman Chemical Supplier No. 3669350 MG
An α2-AR agonist; epidural administration induces antinociception and sedation in donkeys at 50 UG/kg; induces bradycardia in horses from 0.04-0.12 mg/kg