Learn More
Aviva Systems Biology Corp Romo1 Antibody - N-terminal region (ARP58431_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58431_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Romo1. It was validated on Western Blot. . Alternate names: 2010100O12Rik, AI853864, RP23-107C23.2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-474