Learn More
Eurofins Discoverx Ron, active, 10 µg. (14-581)
Supplier:  Eurofins Discoverx 14581
Encompass
N-terminal GST-tagged recombinant, human Ron amino acids 983-end. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-9001