Learn More
Carnabio Usa Inc RON(MST1R) Cytoplasmic domain Wild type Amino Acid 979-1400 NP002438.1 Expressed in Insect (sf21) 100microg
Supplier:  Carnabio Usa Inc 08152100
Encompass
RON(MST1R) Cytoplasmic domain Wild type Amino Acid 979-1400 NP002438.1 Expressed in Insect (sf21) 100microg
Catalog No. 50-252-7806