Learn More
Carnabio Usa Inc RON(MST1R)/10UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 0815210
Encompass
RON(MST1R), Cytoplasmic domain, Wild type, Amino Acid 979-1400, NP_002438.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0974