Learn More
Carnabio Usa Inc RON(MST1R)/5UG/CYTOPLASMIC
Supplier:  Carnabio Usa Inc 081525
Encompass
RON(MST1R), Cytoplasmic domain, Wild type, Amino Acid 979-1400, NP_002438.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0612