Learn More
METTLER TOLEDO 3P INDUSTRIAL RONDO DISPENSING TUBE 1.35 M
Supplier:  METTLER TOLEDO 3P INDUSTRIAL 51108070
Encompass
The Karl Fischer Kit for Rondo 60 transforms a standard Rondo 60 within minutes into a sample changer for Karl Fischer Titrations. The conversion can be accomplished by the user himself and does not need special tools or knowledge. after the conversion, the system allows the automation of up to 20 samples in combination with a Karl Fischer capable DL5x or DL7x titrator. The samples are processed the following way: 1. Samples are filled into the special glass beakers. 2. The beakers are tightly closed with an aluminium lid and a rubber ring. 3. Before titration, the KF head of Rondo 60 pinches through the lid, fills in the methanol. 4. after mixing with the magnetic stirrer, the sample is titrated. The system allows highly precise measurements of water contents down to 100 ppm. The conversion back into a standard Rondo 60 is possible anytime.
Catalog No. NC0068169