Learn More
World Precision Instrument Mini Blumenthal Rongeurs, 11cm, German
SDP
Supplier:  World Precision Instrument 14090G
Encompass
Mini Blumenthal Rongeurs are used for removing bone or other hard tissue during surgical procedures, often in dental procedures. Blumenthal Rongeurs are designed with a pointed, triangular cutting tip for precise cutting and shaping of bone and other hard tissue. These rongeurs have double spring handles, giving you more control. They can be used in a variety of procedures, including orthopedic surgeries, neurosurgery, and plastic surgery. They may be used in studies examining bone growth or regeneration, or in studies exploring the effects of various drugs or treatments on bone tissue. The Mini Blumenthal Rongeurs are smaller than the standard version, and it is for use in more restricted spaces. These surgical rongeurs are available in surgical grade stainless steel or premium German steel. Sterilize them with a chemical sterilant or by autoclaving. To properly care for your surgical instruments, they should be cleaned promptly after every use.
Catalog No. 50-822-202