Learn More
Medchemexpress LLC HY-B0565 500mg Medchemexpress, Ronidazole CAS:7681-76-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0565500MG
Encompass_Preferred
Medchemexpress, HY-B0565 500mg Ronidazole CAS:7681-76-7 Ronidazole is a potent and orally active antiprotozoal and anti-microbial agent. Ronidazole acts as a veterinary agent against Tritrichomonas foetus in cats models. Ronidazole can be used the research of forhistomoniasis and swine dysentery. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2601