Learn More
MSC Ronstan Ball Bearing Swivel, 1670 lb Work Load Limit, 1/4 in Pin Fork/Fork, Stainless Steel
Catalog No. 5000527326
Encompass
This ball bearing swivel is designed for rigging and furling systems, providing smooth articulation and rotation. It features a robust work load limit and is constructed from durable stainless steel, making it suitable for demanding applications.
  • Provides smooth articulation and rotation
  • Work load limit of 1670 lb
  • Overall length of 2.125 in (54.1 mm)
  • Constructed from 316 stainless steel
  • Features a 1/4 in diameter fork/fork pin configuration
Catalog No. 50-005-27326 Supplier MSC Supplier No. 67685321
This ball bearing swivel is designed for rigging and furling systems, providing smooth articulation and rotation. It features a robust work load limit and is constructed from durable stainless steel, making it suitable for demanding applications.
  • Provides smooth articulation and rotation
  • Work load limit of 1670 lb
  • Overall length of 2.125 in (54.1 mm)
  • Constructed from 316 stainless steel
  • Features a 1/4 in diameter fork/fork pin configuration