Learn More
MSC Ronstan Carbine Hook, Stainless Steel 316, 13/32 in Diameter, 4 in Length
Catalog No. 5000531052
Encompass
This carbine hook is manufactured from high-quality stainless steel 316, ensuring durability and corrosion resistance. Designed for quick and secure attachments, it features a 13/32 in diameter and a 4 in overall length, making it suitable for various fastening applications.
  • Made from stainless steel 316
  • 13/32 in diameter
  • 4 in overall length
  • 0.4063 in (10 mm) overall thickness
  • Provides fast and effective means of attachment
Catalog No. 50-005-31052 Supplier MSC Supplier No. 67685602
This carbine hook is manufactured from high-quality stainless steel 316, ensuring durability and corrosion resistance. Designed for quick and secure attachments, it features a 13/32 in diameter and a 4 in overall length, making it suitable for various fastening applications.
  • Made from stainless steel 316
  • 13/32 in diameter
  • 4 in overall length
  • 0.4063 in (10 mm) overall thickness
  • Provides fast and effective means of attachment