Learn More
MSC Ronstan S-Hook, 316 Stainless Steel, 9.5 mm (3/8 in), Closed Eye
Catalog No. 5000529372
Encompass
This S-Hook is constructed from grade 316 stainless steel, ensuring maximum corrosion resistance and durability. It features a 9.5 mm (3/8 in) diameter bar and is designed with a closed eye and a long return leg to provide a positive and secure attachment, effectively preventing accidental release. This electropolished hook is suitable for a wide range of demanding applications.
  • Made from grade 316 stainless steel
  • Features a closed eye and long return leg
  • Ensures positive attachment and avoids accidental release
  • Offers a breaking load capacity of 1760 lb
  • Has a 9.5 mm (3/8 in) diameter bar
  • Electropolished finish for enhanced durability
Catalog No. 50-005-29372 Supplier MSC Supplier No. 02066215
This S-Hook is constructed from grade 316 stainless steel, ensuring maximum corrosion resistance and durability. It features a 9.5 mm (3/8 in) diameter bar and is designed with a closed eye and a long return leg to provide a positive and secure attachment, effectively preventing accidental release. This electropolished hook is suitable for a wide range of demanding applications.
  • Made from grade 316 stainless steel
  • Features a closed eye and long return leg
  • Ensures positive attachment and avoids accidental release
  • Offers a breaking load capacity of 1760 lb
  • Has a 9.5 mm (3/8 in) diameter bar
  • Electropolished finish for enhanced durability