Learn More
MSC Ronstan Snatch Block, Side Release, 2-1/2 in Sheave, 2,200 lb Load Limit
Catalog No. 5000534634
Encompass
The Ronstan Side Release Snatch Block is engineered for versatility and durability in marine environments. Constructed from 316 stainless steel, this block features a 2-1/2 inch sheave and is rated for a 2,200 lb load limit, making it ideal for temporary reefing, sheeting, and general use on boats.
  • Single sheave design for various rigging applications
  • Features a 2-1/2 inch outside diameter sheave for smooth rope handling
  • Constructed from 316 stainless steel for superior corrosion resistance
  • Designed for use with fibrous ropes up to 1/2 inch diameter
  • Work load limit of 1,870 lb
  • Suitable as a temporary reefing block for boats up to 10m (33ft)
  • Functions as a spare or replacement block for general use on larger boats
  • Can be used for temporary sheeting points while changing sheets for boats up to 10m (33ft)
Catalog No. 50-005-34634 Supplier MSC Supplier No. 73524969
The Ronstan Side Release Snatch Block is engineered for versatility and durability in marine environments. Constructed from 316 stainless steel, this block features a 2-1/2 inch sheave and is rated for a 2,200 lb load limit, making it ideal for temporary reefing, sheeting, and general use on boats.
  • Single sheave design for various rigging applications
  • Features a 2-1/2 inch outside diameter sheave for smooth rope handling
  • Constructed from 316 stainless steel for superior corrosion resistance
  • Designed for use with fibrous ropes up to 1/2 inch diameter
  • Work load limit of 1,870 lb
  • Suitable as a temporary reefing block for boats up to 10m (33ft)
  • Functions as a spare or replacement block for general use on larger boats
  • Can be used for temporary sheeting points while changing sheets for boats up to 10m (33ft)