Learn More
Grainger Miller Fusion Roof Anchor Post, Aluminum and Stainless Steel, 310 lb Capacity, 1/Case
Catalog No. 502786146
Encompass_Preferred
The Miller Fusion™ Roof Anchor Post provides a versatile, surface-mount fall protection solution for flat roof environments. Engineered with a unique energy-absorbing load distribution system, this anchor minimizes fall forces on the structure. Its modular design allows for rapid installation on existing roofs without penetrating sub-surface rafters or trusses, supporting both temporary and permanent applications.
  • Features energy-absorbing design that reorients with force
  • Reduces installation time by more than 50%
  • Durable stainless steel and aluminum construction
  • Supports metal sheathing (minimum thickness 24 gauge [0.024 in.])
  • Accommodates trapezoidal spacing from 8 in. (203 mm) to 20 in. (508 mm) in one-inch increments
  • Meets OSHA and ANSI A10.32 and Z359.1-2007 standards
  • Includes rivet kit with sealing washers and mastic tape
Catalog No. 50-278-6146 Supplier Grainger Supplier No. 19D205
The Miller Fusion™ Roof Anchor Post provides a versatile, surface-mount fall protection solution for flat roof environments. Engineered with a unique energy-absorbing load distribution system, this anchor minimizes fall forces on the structure. Its modular design allows for rapid installation on existing roofs without penetrating sub-surface rafters or trusses, supporting both temporary and permanent applications.
  • Features energy-absorbing design that reorients with force
  • Reduces installation time by more than 50%
  • Durable stainless steel and aluminum construction
  • Supports metal sheathing (minimum thickness 24 gauge [0.024 in.])
  • Accommodates trapezoidal spacing from 8 in. (203 mm) to 20 in. (508 mm) in one-inch increments
  • Meets OSHA and ANSI A10.32 and Z359.1-2007 standards
  • Includes rivet kit with sealing washers and mastic tape