The Fusion Roof Anchor Post is a versatile, single-point anchorage connector designed for fall protection during rooftop inspection and maintenance. Engineered to minimize structural impact, this system reorients during a fall to align with the force and activates a patent-pending energy-absorbing system. The anchor attaches to existing roof structures without requiring surface penetration, preserving the integrity of the roofing material.
- Fits standing seam spacing from 11.75 in. (298 mm) to 17 in. (432 mm)
- Features four clamps for secure attachment
- Constructed with zinc-plated and powder-coated steel base and post for environmental durability
- Internal components made of stainless steel to withstand outdoor conditions
- Meets OSHA 1910.66, OSHA 1926.502(d), ANSI A10.32, and ANSI Z359.1 standards
- Maximum capacity of 310 lb.