Learn More
Grainger Condor Roofer's Harness Kit, Universal, With Roofing Anchor, 1 ea
Catalog No. 502783827
Encompass_Preferred
This roofer's harness kit provides a complete fall protection solution for general industry applications. Designed for safety and reliability, the kit includes a universal-fit harness with tongue buckle leg straps and all essential components for secure anchor and lifeline attachment.
  • Includes five-point harness, reusable hinged roof peak anchor, and 50 ft. vertical lifeline
  • Features a self-tracking rope grab with an integral 3 ft. shock-absorbing lanyard
  • Constructed with durable polyester webbing and alloy steel hardware
  • Equipped with back D-ring and subpelvic strap for added security
  • Weight capacity rated for 310 lbs.
  • Packaged in a 5 gal. bucket for convenient storage
  • Meets ANSI Z359.1-2007 standards
Catalog No. 50-278-3827 Supplier Grainger Supplier No. 19F394
This roofer's harness kit provides a complete fall protection solution for general industry applications. Designed for safety and reliability, the kit includes a universal-fit harness with tongue buckle leg straps and all essential components for secure anchor and lifeline attachment.
  • Includes five-point harness, reusable hinged roof peak anchor, and 50 ft. vertical lifeline
  • Features a self-tracking rope grab with an integral 3 ft. shock-absorbing lanyard
  • Constructed with durable polyester webbing and alloy steel hardware
  • Equipped with back D-ring and subpelvic strap for added security
  • Weight capacity rated for 310 lbs.
  • Packaged in a 5 gal. bucket for convenient storage
  • Meets ANSI Z359.1-2007 standards