Learn More
Euthanex Room Air Pump to be used as an alternate to O2 Tanks for Anesthesia Systems. Max. 10 LPM, 2.2 PSI.
Catalog No. NC2259359
Encompass_Preferred
The EZ-AP122 Room Air Pump can be used with the Classic and Single Animal series anesthesia systems to drive air through the vaporizer. The EZ-AP122 is a quiet and economical alternative to oxygen tanks. Features an inlet filter and includes an outlet tube with a DISS female fitting. Therefore is easily connects to an anesthesia machine. It can be connected directly to the inlet of the system and controlled using the flow meter on the anesthesia machine.
Catalog No. NC2259359 Supplier Euthanex Supplier No. EZAP122
The EZ-AP122 Room Air Pump can be used with the Classic and Single Animal series anesthesia systems to drive air through the vaporizer. The EZ-AP122 is a quiet and economical alternative to oxygen tanks. Features an inlet filter and includes an outlet tube with a DISS female fitting. Therefore is easily connects to an anesthesia machine. It can be connected directly to the inlet of the system and controlled using the flow meter on the anesthesia machine.