Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mystaire™ Room Air Purifier
SDP

Catalog No. 15338152 Shop All Mystaire Products
Change view
Click to view available options
Type:
Dual-Replacement Capture Arm
Fume Extractor
Single-Replacement Capture Arm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type
15-338-152 Dual-Replacement Capture Arm
15-338-166 Fume Extractor
15-338-151 Single-Replacement Capture Arm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 15-338-152 Supplier Mystaire™ Supplier No. FE1215

Target fumes at the source or eliminate room air quality issues.

  • Sparkless and brushless centrifugal fan for quiet operation
  • Epoxy coated aluminum construction
  • HEPA and/or Carbon Filtration
  • HEPA or carbon filter depending on application
  • Lightweight and portable
  • Fume Extractor: One or two adjustable capture arms for easy movement to the source of contamination

Specifications

For Use With (Equipment) Mystaire Ductless Workstation
Type Dual-Replacement Capture Arm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.