Description
- Sparkless and brushless centrifugal fan for quiet operation
- Epoxy coated aluminum construction
- HEPA and/or Carbon Filtration
- HEPA or carbon filter depending on application
- Lightweight and portable
- Fume Extractor: One or two adjustable capture arms for easy movement to the source of contamination
Specifications
Specifications
| For Use With (Equipment) | Mystaire Ductless Workstation |
| Type | Fume Extractor |