Learn More
Johnstone Supply ROOM TMP RCRDNG CHRT PPR 60/PK
Catalog No. NC9958828
Encompass_Preferred
12 recorders in 1. For refrigerator, freezer and environmental temperature measurement. Measures and records temperatures in air, gas, liquids, powders and semi-solids. Temperature ranges of: -40�F to +30�F, -20�F to +50�F or 50�F to 120�F. Recording speeds of 6 hours, 24 hours, 7 days and 31 days. Selectable, single turn or continuous chart rotation. Remote reading 4" long submersible stainless steel probe with 15' sensor wire. Standard 120V operation/special 12V feature allows for use in vehicular or marine operation. Battery backup. Includes 60 assorted, 6" diameter charts and 2 pens
Catalog No. NC9958828 Supplier Johnstone Supply Supplier No. CR8718
12 recorders in 1. For refrigerator, freezer and environmental temperature measurement. Measures and records temperatures in air, gas, liquids, powders and semi-solids. Temperature ranges of: -40�F to +30�F, -20�F to +50�F or 50�F to 120�F. Recording speeds of 6 hours, 24 hours, 7 days and 31 days. Selectable, single turn or continuous chart rotation. Remote reading 4" long submersible stainless steel probe with 15' sensor wire. Standard 120V operation/special 12V feature allows for use in vehicular or marine operation. Battery backup. Includes 60 assorted, 6" diameter charts and 2 pens