Learn More
Roosterbio Inc RoosterNourish™-MSC: Traditional bioprocess medium for the expansion of hMSCs
SDP
Catalog No. NC2204744
Encompass
RoosterNourish™-MSC is a translation-friendly, bioprocess medium designed to eliminate media exchanges during rapid expansion of human mesenchymal stem/stromal cells (hMSCs) from multiple tissue sources. This medium supports in vitro expansion of hMSCs in a 2D batch process.  RoosterNourish™-MSC has unparalleled productivity (Millions of cells produced per Liters of media consumed) compared to other MSC media on the market. Used alongside our RoosterVial™-MSC and following RoosterBio protocols, we guarantee 10X hMSC expansion within 4-6 days. This medium has been used to generate high volumes of hMSC critical raw materials worldwide and has supported top regenerative medicine research, product developers, and translational institutions.Intended Use: Research Use Only
This item is not returnable. View return policy
Catalog No. NC2204744 Supplier Roosterbio Inc Supplier No. K82003
RoosterNourish™-MSC is a translation-friendly, bioprocess medium designed to eliminate media exchanges during rapid expansion of human mesenchymal stem/stromal cells (hMSCs) from multiple tissue sources. This medium supports in vitro expansion of hMSCs in a 2D batch process.  RoosterNourish™-MSC has unparalleled productivity (Millions of cells produced per Liters of media consumed) compared to other MSC media on the market. Used alongside our RoosterVial™-MSC and following RoosterBio protocols, we guarantee 10X hMSC expansion within 4-6 days. This medium has been used to generate high volumes of hMSC critical raw materials worldwide and has supported top regenerative medicine research, product developers, and translational institutions.Intended Use: Research Use Only