Learn More
Roosterbio Inc RoosterCollect EV Pro™: A media kit designed for post-cell expansion EV production and collection.
SDP
Catalog No. NC2589109
Encompass
RoosterCollect-EV Pro™ is a translation-friendly, low particulate, chemically defined Extracellular Vesicle (EV) collection media kit designed to increase total hMSC-EV yields.  For use post-cell expansion, this medium supports cell health during hMSC-based EV collection to elongate the collection window, increase EV yields, and reduce variability in 3D Bioreactor as well as 2D flask culture systems. As part of RoosterBio’s EV Production System – a bioprocess engine for rapid EV production – each RoosterCollect EV Pro kit supports RoosterBio’s scalable and translation-friendly Development Grade xeno-free (XF) product portfolio, including high volume hMSCs and bioprocess media. Across tissue types, this low-particulate medium and paired  2D batch or 3D bioreactor fed-batch bioprocess scales with your stage of product development – ensuring unparalleled hMSC-EV production. Industrialize your hMSC-EV supply chain with a complete extracellular vesicle (EV) production workflow. 
Catalog No. NC2589109 Supplier Roosterbio Inc Supplier No. K41001
RoosterCollect-EV Pro™ is a translation-friendly, low particulate, chemically defined Extracellular Vesicle (EV) collection media kit designed to increase total hMSC-EV yields.  For use post-cell expansion, this medium supports cell health during hMSC-based EV collection to elongate the collection window, increase EV yields, and reduce variability in 3D Bioreactor as well as 2D flask culture systems. As part of RoosterBio’s EV Production System – a bioprocess engine for rapid EV production – each RoosterCollect EV Pro kit supports RoosterBio’s scalable and translation-friendly Development Grade xeno-free (XF) product portfolio, including high volume hMSCs and bioprocess media. Across tissue types, this low-particulate medium and paired  2D batch or 3D bioreactor fed-batch bioprocess scales with your stage of product development – ensuring unparalleled hMSC-EV production. Industrialize your hMSC-EV supply chain with a complete extracellular vesicle (EV) production workflow. 