Learn More
Roosterbio Inc RoosterCollect™-EV: Collection Medium for Extracellular Vesicles (EVs)
SDP
Catalog No. NC2589107
Encompass
RoosterCollect™-EV is a chemically defined extracellular vesicle (EV) collection media designed to seamlessly integrate into EV, exosome, or cell secretome production platforms. Designed for EV manufacturing across many cell and tissue types in both 2D planar and 3D bioreactor culture conditions, this protein-free and low-particulate media enables “clean collection” resulting in higher yields and easier downstream processing. RoosterCollect™-EV is easily integrated into existing workflows to support EV collection from preferred cell types, or when combined with RoosterBio’s hMSCs and expansion media, RoosterCollect™-EV turns the RoosterBio cell culture platform into a highly optimized extracellular vesicle production platform.
Catalog No. NC2589107 Supplier Roosterbio Inc Supplier No. M2001
RoosterCollect™-EV is a chemically defined extracellular vesicle (EV) collection media designed to seamlessly integrate into EV, exosome, or cell secretome production platforms. Designed for EV manufacturing across many cell and tissue types in both 2D planar and 3D bioreactor culture conditions, this protein-free and low-particulate media enables “clean collection” resulting in higher yields and easier downstream processing. RoosterCollect™-EV is easily integrated into existing workflows to support EV collection from preferred cell types, or when combined with RoosterBio’s hMSCs and expansion media, RoosterCollect™-EV turns the RoosterBio cell culture platform into a highly optimized extracellular vesicle production platform.