Learn More
Roosterbio Inc RoosterGEM™: Complete Genetic Engineering Medium for Viral & Non-Viral Applications
SDP
Supplier:  Roosterbio Inc M40200
Encompass
Engineered from the ground up, RoosterGEM are complete media built to solve key challenges in both viral and non-viral MSC and primary cell engineering. Traditionally, genetic modification of primary cells has low efficiency of transfer requiring increased viral  and non-viral agent costs, with significant time and resources allocated to media and process optimization. RoosterGEM is a highly efficient Complete Genetic Engineering Medium, formulated to increase genetic engineering efficiencies, enabling a reduction in key raw material costs and more readily meet regulatory guidelines. When paired with RoosterBio’s industrialized supply chain of high-volume Xeno-free hMSCs, ultra-productive expansion media, and cGMP-compatible processes – you enlist an end-to-end Genetic Engineering MSC-platform to streamline your transient or integrated gene transfer, de-risk your product & process development, and streamline your path to clinical translation.
Catalog No. NC2589113