Learn More
Roosterbio Inc RoosterNourish™-MSC-XF: A standardized, xeno-free bioprocess medium for the rapid expansion of hMSCs
SDP
Catalog No. NC2208560
Encompass
RoosterNourish™-MSC-XF is a translation-friendly, xeno-free (XF) bioprocess medium designed to eliminate media exchanges during rapid expansion of human mesenchymal stem/stromal cells (hMSCs) from multiple tissue sources. This medium supports in vitro expansion of hMSCs in a 2D batch process or in a 3D bioreactor fed-batch process with our RoosterReplenish™-MSC-XF bioreactor feed.  RoosterNourish™-MSC-XF has unparalleled productivity (Millions of cells produced per Liters of media consumed) compared to other MSC media on the market. Used alongside our XF RoosterVial™-MSC and following RoosterBio protocols, we guarantee 10X hMSC expansion within 4-6 days. This medium has been used to generate high volumes of hMSC critical raw materials worldwide and has supported top regenerative medicine research, product developers, and translational institutions.Intended Use: Research Use Only
This item is not returnable. View return policy
Catalog No. NC2208560 Supplier Roosterbio Inc Supplier No. K82016
RoosterNourish™-MSC-XF is a translation-friendly, xeno-free (XF) bioprocess medium designed to eliminate media exchanges during rapid expansion of human mesenchymal stem/stromal cells (hMSCs) from multiple tissue sources. This medium supports in vitro expansion of hMSCs in a 2D batch process or in a 3D bioreactor fed-batch process with our RoosterReplenish™-MSC-XF bioreactor feed.  RoosterNourish™-MSC-XF has unparalleled productivity (Millions of cells produced per Liters of media consumed) compared to other MSC media on the market. Used alongside our XF RoosterVial™-MSC and following RoosterBio protocols, we guarantee 10X hMSC expansion within 4-6 days. This medium has been used to generate high volumes of hMSC critical raw materials worldwide and has supported top regenerative medicine research, product developers, and translational institutions.Intended Use: Research Use Only