Learn More
Roosterbio Inc RoosterVial™-hBM: Human Bone Marrow-Derived Mesenchymal Stem/Stromal Cells
SDP
Supplier:  Roosterbio Inc MSC001
Encompass
RoosterVial™-hBM are Human Bone-Marrow Derived Mesenchymal Stem/Stromal Cells (hBM-MSCs) isolated from adult human iliac crest from a variety of normal, healthy donors and expanded to a population doubling level (PDL) of 6-10, post-mononuclear cell isolation. Available in both 1M or high-volume 10M cell per vial formats to meet your needs throughout product development.Each lot is release tested to expand >10-fold within 4 to 6 days in a media exchange free process when paired with RoosterNourishTM-MSC bioprocess medium and conforms to ISCT minimal criteria for hMSC identity giving added confidence in quality, reproducibility, and ease of integration.Designed for robust, reproducible, standardized scale-up and supported by our media exchange free 2D and 3D bioreactor processes and RoosterNourish™-MSC expansion media, RoosterVial™-hBM can efficiently and reproducibly get you the millions or billions of hMSCs to accelerate your Research and Product Development.
Catalog No. NC2589080