Learn More
Roosterbio Inc Xeno-Free RoosterVial™-hUC: Xeno-Free Human Umbilical Cord-Derived Mesenchymal Stem/Stromal Cells
SDP
Catalog No. NC2589087
Encompass
RoosterVial™-hUC-XF are Human Umbilical Cord Derived Mesenchymal Stem/Stromal Cells (hUC-MSCs) isolated from the perivascular Wharton’s Jelly region of the human umbilical cord and expanded to a population doubling level (PDL) of 14-20.  Available in both 1M or high-volume 10M cell per vial formats to meet your needs throughout product development. Each lot is release tested to expand >10-fold within 4 to 6 days in a media exchange free process when paired with RoosterNourish™-MSC-XF bioprocess medium and conforms to ISCT minimal criteria for hMSC identity. Accompanying QC brief gives added confidence in quality, reproducibility, and ease of integration. Designed for robust, reproducible, standardized XF scale-up and supported by our media exchange free 2D and 3D bioreactor processes and RoosterNourish™-MSC-XF expansion media, XF RoosterVial™-hUC can efficiently and reproducibly get you the millions or billions of hMSCs to accelerate your
Catalog No. NC2589087 Supplier Roosterbio Inc Supplier No. C43001UC
RoosterVial™-hUC-XF are Human Umbilical Cord Derived Mesenchymal Stem/Stromal Cells (hUC-MSCs) isolated from the perivascular Wharton’s Jelly region of the human umbilical cord and expanded to a population doubling level (PDL) of 14-20.  Available in both 1M or high-volume 10M cell per vial formats to meet your needs throughout product development. Each lot is release tested to expand >10-fold within 4 to 6 days in a media exchange free process when paired with RoosterNourish™-MSC-XF bioprocess medium and conforms to ISCT minimal criteria for hMSC identity. Accompanying QC brief gives added confidence in quality, reproducibility, and ease of integration. Designed for robust, reproducible, standardized XF scale-up and supported by our media exchange free 2D and 3D bioreactor processes and RoosterNourish™-MSC-XF expansion media, XF RoosterVial™-hUC can efficiently and reproducibly get you the millions or billions of hMSCs to accelerate your