Learn More
Med Vet International Root Tip Pick, J0042X
Catalog No. 5000585383
Encompass_Preferred
Heid Brink tooth ejector. Sometimes during extraction, tooth fragments can break off and remain in the socket. This instrument has a thin delicate tip that helps loosen and remove those remaining fragments.
Catalog No. 50-005-85383 Supplier Med Vet International Supplier No. JOR42X
Heid Brink tooth ejector. Sometimes during extraction, tooth fragments can break off and remain in the socket. This instrument has a thin delicate tip that helps loosen and remove those remaining fragments.