Learn More
Roots and Stems Prepared Slides
SCI_ED
Catalog No. p-2379480
Change view
Click to view available options
Type:
Allium cepa (Onion Liliaceae); Root tip squash preparation for mitosis, Feulgen stain
Catalog No. Type
S10663 Allium cepa (Onion Liliaceae); Root tip squash preparation for mitosis, Feulgen stain
1 options
This item is not returnable. View return policy
Catalog No. S10663 Supplier Triarch Inc Supplier No. 142F
This item is not returnable. View return policy
Slides give magnified view of Gymnosperms and Angiosperms for detailed study.
  • Pine cross section provides excellent view of secondary xylem for discussing formation of annual rings
  • Determine the difference between monocot and dicot roots, stems, and leaves
  • Flower reproductive parts such as pollen and pollen tubes are clearly visible
Specifications
Type Allium cepa (Onion Liliaceae); Root tip squash preparation for mitosis, Feulgen stain
Specimen Type Roots and Stems
Slide Type Prepared Slide