Learn More
Medchemexpress LLC Ropidoxuridine | 93265-81-7 | MFCD13194773 | 99.6% | 338.10 g/mol | C9H11IN2O4 | 5 MG
SDP
Catalog No. 5000222158
Encompass_Preferred
Ropidoxuridine (IPdR) is an orally available halogenated thymidine analog investigated as a potential radiosensitizer for use in human tumors. Supplied for research use, the compound is available as a solid or as a DMSO solution and is characterized by high purity and defined storage recommendations.
  • Orally available halogenated thymidine analog.
  • Investigated as a potential radiosensitizer for human tumors.
  • Available as solid in multiple milligram sizes and as a 10 mM solution in DMSO.
  • High purity (99.59%).
  • Physical appearance: light yellow to green-yellow solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-22158 Supplier Medchemexpress LLC Supplier No. HY137425MG
Ropidoxuridine (IPdR) is an orally available halogenated thymidine analog investigated as a potential radiosensitizer for use in human tumors. Supplied for research use, the compound is available as a solid or as a DMSO solution and is characterized by high purity and defined storage recommendations.
  • Orally available halogenated thymidine analog.
  • Investigated as a potential radiosensitizer for human tumors.
  • Available as solid in multiple milligram sizes and as a 10 mM solution in DMSO.
  • High purity (99.59%).
  • Physical appearance: light yellow to green-yellow solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).