Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ropinirole hydrochloride | 91374-20-8 | 99.9% | 50 MG
SDP

Catalog No. 5000405901
Encompass_Preferred

Ropinirole hydrochloride is an orally active, potent D3/D2 receptor agonist with a Ki of 29 nM for the D2 receptor. It exhibits pEC50s of 7.4, 8.4, and 6.8 for hD2, hD3, and hD4 receptors, respectively, and demonstrates no affinity for D1 receptors. This compound is under investigation for its therapeutic potential in treating Parkinson's disease.

  • Potent D3/D2 receptor agonist
  • Ki of 29 nM for D2 receptor
  • pEC50s of 7.4 (hD2), 8.4 (hD3), and 6.8 (hD4)
  • No affinity for D1 receptors
  • Higher affinity for D3 receptors compared to D2 and D4 receptors
  • Weakly active at alpha 2-adrenoceptors and 5-HT2 receptors
  • Inactive at 5-HT1, benzodiazepine, gamma-aminobutyric acid receptors, or alpha 1 and beta-adrenoceptors
  • Investigated for potential in treating Parkinson's disease

Catalog No. 50-004-05901 Supplier Medchemexpress LLC Supplier No. HYB0623A50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ropinirole hydrochloride is an orally active, potent D3/D2 receptor agonist with a Ki of 29 nM for the D2 receptor. It exhibits pEC50s of 7.4, 8.4, and 6.8 for hD2, hD3, and hD4 receptors, respectively, and demonstrates no affinity for D1 receptors. This compound is under investigation for its therapeutic potential in treating Parkinson's disease.

  • Potent D3/D2 receptor agonist
  • Ki of 29 nM for D2 receptor
  • pEC50s of 7.4 (hD2), 8.4 (hD3), and 6.8 (hD4)
  • No affinity for D1 receptors
  • Higher affinity for D3 receptors compared to D2 and D4 receptors
  • Weakly active at alpha 2-adrenoceptors and 5-HT2 receptors
  • Inactive at 5-HT1, benzodiazepine, gamma-aminobutyric acid receptors, or alpha 1 and beta-adrenoceptors
  • Investigated for potential in treating Parkinson's disease

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.