Learn More
Med Vet International RX Ropivacaine HCL Injection (Generic Naropin), 0.5%, 30mL, 10/Pack
Catalog No. 503042984
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Ropivacaine HCL Injection contains 150mg/30ml. This product should be stored at 20° to 25°c (68° to 77°F). Do not use this product if the solution is discolored or contains precipitate. This product is not for intravenous administration. The included vial stopper is not made with natural rubber latex.
Catalog No. 50-304-2984 Supplier Med Vet International Supplier No. RX43066002310
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Ropivacaine HCL Injection contains 150mg/30ml. This product should be stored at 20° to 25°c (68° to 77°F). Do not use this product if the solution is discolored or contains precipitate. This product is not for intravenous administration. The included vial stopper is not made with natural rubber latex.