Learn More
Medchemexpress LLC Ropivacaine | 84057-95-4 | 100.0% | 5 MG
SDP
Catalog No. 5000409461
Encompass_Preferred
Ropivacaine is a potent sodium channel blocker that blocks impulse conduction by reversibly inhibiting sodium ion influx in nerve fibers. It also acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1. It is utilized in the research of neuropathic pain management.
  • Blocks impulse conduction by reversibly inhibiting sodium ion influx in nerve fibers.
  • Acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane.
  • Utilized in the research of neuropathic pain management.
Catalog No. 50-004-09461 Supplier Medchemexpress LLC Supplier No. HYB05635MG
Ropivacaine is a potent sodium channel blocker that blocks impulse conduction by reversibly inhibiting sodium ion influx in nerve fibers. It also acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1. It is utilized in the research of neuropathic pain management.
  • Blocks impulse conduction by reversibly inhibiting sodium ion influx in nerve fibers.
  • Acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane.
  • Utilized in the research of neuropathic pain management.