Learn More
Medchemexpress LLC Ropivacaine | 84057-95-4 | MFCD00078774 | 100.0% | 50 MG
SDP
Catalog No. 5000408689
Encompass_Preferred
Overview of Ropivacaine
Ropivacaine is a potent sodium channel blocker. It works by blocking impulse conduction through reversible inhibition of sodium ion influx in nerve fibers. Additionally, Ropivacaine acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. It is utilized in the research of neuropathic pain management.
Features and Benefits
  • Potent sodium channel blocker.
  • Blocks impulse conduction by inhibiting sodium ion influx.
  • Inhibits K2P TREK-1 (IC50 of 402.7 μM).
  • Used for neuropathic pain management research.
  • Epidural administration blocks neuropathic pain and delays its development.
  • Inhibits pressure-induced increases in filtration coefficient.
  • Prevents pressure-induced lung edema and hyperpermeability.
  • Inhibits pressure-induced NO production.
Catalog No. 50-004-08689 Supplier Medchemexpress LLC Supplier No. HYB056350MG
Overview of Ropivacaine
Ropivacaine is a potent sodium channel blocker. It works by blocking impulse conduction through reversible inhibition of sodium ion influx in nerve fibers. Additionally, Ropivacaine acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. It is utilized in the research of neuropathic pain management.
Features and Benefits
  • Potent sodium channel blocker.
  • Blocks impulse conduction by inhibiting sodium ion influx.
  • Inhibits K2P TREK-1 (IC50 of 402.7 μM).
  • Used for neuropathic pain management research.
  • Epidural administration blocks neuropathic pain and delays its development.
  • Inhibits pressure-induced increases in filtration coefficient.
  • Prevents pressure-induced lung edema and hyperpermeability.
  • Inhibits pressure-induced NO production.