Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ropivacaine-d7 | 684647-62-9 | 99.4% | 281.44 g·mol⁻1 | C17H19D7N2O | 10 MG
SDP

Catalog No. 5000298936
Encompass_Preferred

Ropivacaine-d7 is a deuterium-labeled analog of ropivacaine supplied as a solid analytical standard for use in quantitative and preclinical research, including mass spectrometry and pharmacokinetic studies. It is intended as an internal standard for assays investigating sodium channel activity and neuropathic pain models.

  • Deuterium-labeled internal standard for quantitative LC-MS assays.
  • High isotopic and chemical purity (99.4%).
  • Molecular weight 281.44 g·mol⁻1 and formula C17H19D7N2O.
  • Available in small packaged quantities suitable for analytical use (10 MG).
  • Solid form for stable storage and handling.

Catalog No. 50-002-98936 Supplier Medchemexpress LLC Supplier No. HYB0563S110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ropivacaine-d7 is a deuterium-labeled analog of ropivacaine supplied as a solid analytical standard for use in quantitative and preclinical research, including mass spectrometry and pharmacokinetic studies. It is intended as an internal standard for assays investigating sodium channel activity and neuropathic pain models.

  • Deuterium-labeled internal standard for quantitative LC-MS assays.
  • High isotopic and chemical purity (99.4%).
  • Molecular weight 281.44 g·mol⁻1 and formula C17H19D7N2O.
  • Available in small packaged quantities suitable for analytical use (10 MG).
  • Solid form for stable storage and handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.