Learn More
Medchemexpress LLC Ropivacaine-d7 | 684647-62-9 | 99.69% | 1 MG
SDP
Catalog No. 5000408375
Encompass_Preferred
Ropivacaine-d7 is a deuterium labeled Ropivacaine. Ropivacaine is a potent sodium channel blocker, inhibiting impulse conduction by reversibly blocking sodium ion influx in nerve fibers. It also acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell membranes. Ropivacaine is used for the research of neuropathic pain management, and is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-08375 Supplier Medchemexpress LLC Supplier No. HYB0563S11MG
Ropivacaine-d7 is a deuterium labeled Ropivacaine. Ropivacaine is a potent sodium channel blocker, inhibiting impulse conduction by reversibly blocking sodium ion influx in nerve fibers. It also acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell membranes. Ropivacaine is used for the research of neuropathic pain management, and is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.