Learn More
Apexbio Technology LLC Ropivacaine HCl 98717-15-8 10mM (in 1mL H2O)
SDP
Catalog No. 5000062001
Encompass_Preferred
Ropivacaine HCl (CAS 98717-15-8) is a local anesthetic agent commonly employed in biomedical research to examine neuronal function and signal transmission Mechanistically ropivacaine hydrochloride acts by reversibly blocking the influx of sodium ions across neuronal membranes thereby inhibiting the conduction of nerve impulses Due to this mode of action it serves as a useful pharmacological tool in studies designed to elucidate pain signaling pathways sodium channel behavior and peripheral nerve function
Catalog No. 50-000-62001 Supplier Apexbio Technology LLC Supplier No. B20235.1
Ropivacaine HCl (CAS 98717-15-8) is a local anesthetic agent commonly employed in biomedical research to examine neuronal function and signal transmission Mechanistically ropivacaine hydrochloride acts by reversibly blocking the influx of sodium ions across neuronal membranes thereby inhibiting the conduction of nerve impulses Due to this mode of action it serves as a useful pharmacological tool in studies designed to elucidate pain signaling pathways sodium channel behavior and peripheral nerve function