Learn More
Medchemexpress LLC Ropivacaine (hydrochloride) | 98717-15-8 | 99.5% | 1 ML
SDP
Catalog No. 5000409009
Encompass_Preferred
Ropivacaine hydrochloride is a potent sodium channel blocker that reversibly inhibits sodium ion influx in nerve fibers to block impulse conduction. It also acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. It is widely used for neuropathic pain management in vivo.
  • Potent sodium channel blocker
  • Reversibly inhibits sodium ion influx
  • Inhibits K2P TREK-1 with an IC50 of 402.7 μM
  • Used for neuropathic pain management in vivo
  • Effectively blocks neuropathic pain via epidural administration
  • Prevents pressure-induced lung edema and associated hyperpermeability
Catalog No. 50-004-09009 Supplier Medchemexpress LLC Supplier No. HYB0563B10MM/1ML
Add to Cart
Add to Cart
Ropivacaine hydrochloride is a potent sodium channel blocker that reversibly inhibits sodium ion influx in nerve fibers to block impulse conduction. It also acts as an inhibitor of K2P (two-pore domain potassium channel) TREK-1 with an IC50 of 402.7 μM in COS-7 cell's membrane. It is widely used for neuropathic pain management in vivo.
  • Potent sodium channel blocker
  • Reversibly inhibits sodium ion influx
  • Inhibits K2P TREK-1 with an IC50 of 402.7 μM
  • Used for neuropathic pain management in vivo
  • Effectively blocks neuropathic pain via epidural administration
  • Prevents pressure-induced lung edema and associated hyperpermeability