Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Ropivacaine (hydrochloride) | 98717-15-8 | 99.5% | 50 MG
SDP

Catalog No. 5000407849
Encompass_Preferred

Ropivacaine hydrochloride is a chemical compound primarily used for laboratory research and the manufacturing of other substances. It presents as a white to off-white solid, with a melting point ranging from 260-262°C, and is stable when stored under recommended conditions.

  • Molecular formula: C17H27ClN2O
  • Molecular weight: 310.86
  • CAS number: 98717-15-8
  • Appearance: White to off-white solid
  • Purity (LCMS): 99.5%
  • Recommended storage: 4°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Identified uses: Laboratory chemicals and substance manufacturing
  • Shipping conditions: Room temperature if less than 2 weeks
  • Avoid strong acids/alkalis, strong oxidising/reducing agents
  • For research use only

Catalog No. 50-004-07849 Supplier Medchemexpress LLC Supplier No. HYB0563B50MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Ropivacaine hydrochloride is a chemical compound primarily used for laboratory research and the manufacturing of other substances. It presents as a white to off-white solid, with a melting point ranging from 260-262°C, and is stable when stored under recommended conditions.

  • Molecular formula: C17H27ClN2O
  • Molecular weight: 310.86
  • CAS number: 98717-15-8
  • Appearance: White to off-white solid
  • Purity (LCMS): 99.5%
  • Recommended storage: 4°C, sealed, away from moisture
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
  • Identified uses: Laboratory chemicals and substance manufacturing
  • Shipping conditions: Room temperature if less than 2 weeks
  • Avoid strong acids/alkalis, strong oxidising/reducing agents
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.