Learn More
Apexbio Technology LLC Ropivacaine hydrochloride monohydrate 132112-35-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000096447
Encompass_Preferred
Ropivacaine hydrochloride monohydrate (CAS 132112-35-7) is a long-acting amide local anesthetic widely studied for its pharmacological and electrophysiological properties It inhibits ATP-sensitive potassium (KATP) channels particularly SUR2A/Kir6 2 as demonstrated in COS-7 cell models where inhibition is concentration-dependent and reversible Reported IC50 values for SUR2A/Kir6 2 SUR2B/Kir6 1 SUR2B/Kir6 2 and Kir6 2 C36 channels are 249 11 2235 115 2442 132 and 2375 179 M respectively Additionally ropivacaine hydrochloride monohydrate suppresses sarcolemmal KATP channels in cardiovascular tissues via stereoselective and tissue-specific mechanisms supporting its utility in studies of ion channel pharmacology and cardiovascular safety profiles
Catalog No. 50-000-96447 Supplier Apexbio Technology LLC Supplier No. B33775.1
Ropivacaine hydrochloride monohydrate (CAS 132112-35-7) is a long-acting amide local anesthetic widely studied for its pharmacological and electrophysiological properties It inhibits ATP-sensitive potassium (KATP) channels particularly SUR2A/Kir6 2 as demonstrated in COS-7 cell models where inhibition is concentration-dependent and reversible Reported IC50 values for SUR2A/Kir6 2 SUR2B/Kir6 1 SUR2B/Kir6 2 and Kir6 2 C36 channels are 249 11 2235 115 2442 132 and 2375 179 M respectively Additionally ropivacaine hydrochloride monohydrate suppresses sarcolemmal KATP channels in cardiovascular tissues via stereoselective and tissue-specific mechanisms supporting its utility in studies of ion channel pharmacology and cardiovascular safety profiles